4-Benzylcarbamoyl-3-chlorophenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BB-2761-1G BB-2761-5G
COBBBB-2761-1GEA 0 GBP
COBBBB-2761-1G COBBBB-2761-5G
4-Benzylcarbamoyl-3-chlorophenylboronic acid
4-Benzylcarbamoyl-3-chlorophenylboronic acid
Formula: C₁₄H₁₃BClNO₃
MDL Number: MFCD07363769
CAS Number: 850589-42-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR