4-Biphenylboronic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: [1,1'-Biphenyl]-4-ylboronic acid

021833-25G 021833-5G
FLUO021833-25GEA 0 GBP
FLUO021833-25G FLUO021833-5G
4-Biphenylboronic acid
4-Biphenylboronic acid
Formula: C₁₂H₁₁BO₂
MW: 198.03 g/mol
MDL Number: MFCD00093311
CAS Number: 5122-94-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR