4-Biphenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: [1,1'-Biphenyl]-4-ylboronic acid

BB-2236-5G BB-2236-25G BB-2236-100G
COBBBB-2236-5GEA 0 GBP
COBBBB-2236-5G COBBBB-2236-25G COBBBB-2236-100G
4-Biphenylboronic acid
4-Biphenylboronic acid
Formula: C₁₂H₁₁BO₂
MDL Number: MFCD00093311
CAS Number: 5122-94-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR