4-Biphenylboronic acid pinacol ester
Supplier: COMBI-BLOCKS INC.
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
COBBPN-2236-5GEA
0
GBP
COBBPN-2236-5G
COBBPN-2236-1G
4-Biphenylboronic acid pinacol ester
4-Biphenylboronic acid pinacol ester
Formula:
C₁₈H₂₁BO₂ MW: 280.17 g/mol |
MDL Number:
MFCD07368291 CAS Number: 144432-80-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...