3'-Amino-2'-hydroxy-5'-methylacetophenone

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

011346-5G 011346-1G
FLUO011346-5GEA 0 GBP
FLUO011346-5G FLUO011346-1G
3'-Amino-2'-hydroxy-5'-methylacetophenone
3'-Amino-2'-hydroxy-5'-methylacetophenone
Formula: C₉H₁₁NO₂
MDL Number: MFCD09996884
CAS Number: 70977-71-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR