2-Nitro-4-(trifluoromethyl)benzyl bromide
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
010344-1G
010344-5G
FLUO010344-1GEA
0
GBP
FLUO010344-1G
FLUO010344-5G
2-Nitro-4-(trifluoromethyl)benzyl bromide
2-Nitro-4-(trifluoromethyl)benzyl bromide
Formula:
C₈H₅BrF₃NO₂ |
MDL Number:
MFCD03095413 CAS Number: 162333-02-0 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...