2-Bromo-4'-(trifluoromethyl)acetophenone

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

011236-1G 011236-5G 011236-25G
FLUO011236-1GEA 0 GBP
FLUO011236-1G FLUO011236-5G FLUO011236-25G
2-Bromo-4'-(trifluoromethyl)acetophenone
2-Bromo-4'-(trifluoromethyl)acetophenone
Formula: C₉H₆BrF₃O
MDL Number: MFCD00126489
CAS Number: 383-53-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR