2-(Phenylsulphonyl)acetophenone

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

001322-25G 001322-100G
FLUO001322-25GEA 0 GBP
FLUO001322-25G FLUO001322-100G
2-(Phenylsulphonyl)acetophenone
2-(Phenylsulphonyl)acetophenone
Formula: C₁₄H₁₂O₃S
MDL Number: MFCD00025043
CAS Number: 3406-03-9
EINECS: 222-292-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR