3-Bromo-2-chloro-5-(trifluoromethyl)phenylboronic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BB-5020-5G
BB-5020-1G
COBBBB-5020-5GEA
0
GBP
COBBBB-5020-5G
COBBBB-5020-1G
3-Bromo-2-chloro-5-(trifluoromethyl)phenylboronic acid
3-Bromo-2-chloro-5-(trifluoromethyl)phenylboronic acid
Formula:
C₇H₄BBrClF₃O₂ |
MDL Number:
MFCD09800879 CAS Number: 957120-85-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...