4-(2-Carboxyethyl)phenylboronic acid pinacol ester

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)propanoic acid

PN-2055-5G PN-2055-1G
COBBPN-2055-5GEA 0 GBP
COBBPN-2055-5G COBBPN-2055-1G
4-(2-Carboxyethyl)phenylboronic acid pinacol ester
4-(2-Carboxyethyl)phenylboronic acid pinacol ester
Formula: C₁₅H₂₁BO₄
MW: 276.1400 g/mol
MDL Number: MFCD11053827
CAS Number: 797755-11-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR