3-(Carboxymethyl)phenylboronic acid pinacol ester

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PN-3907-5G PN-3907-1G
COBBPN-3907-5GEA 0 GBP
COBBPN-3907-5G COBBPN-3907-1G
3-(Carboxymethyl)phenylboronic acid pinacol ester
3-(Carboxymethyl)phenylboronic acid pinacol ester
Formula: C₁₄H₁₉BO₄
MDL Number: MFCD04973389
CAS Number: 79775-05-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR