N-Fluorobenzenesulphonimide

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

006185-5G 006185-10G 006185-50G 006185-100G 006185-25G
FLUO006185-5GEA 0 GBP
FLUO006185-5G FLUO006185-10G FLUO006185-50G FLUO006185-100G FLUO006185-25G
N-Fluorobenzenesulphonimide
N-Fluorobenzenesulphonimide
Formula: C₁₂H₁₀FNO₄S₂
MW: 315.34 g/mol
MDL Number: MFCD00144885
CAS Number: 133745-75-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR