4-(Methylsulphinyl)phenol

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OT-0402-5G OT-0402-25G OT-0402-1G
COBBOT-0402-5GEA 0 GBP
COBBOT-0402-5G COBBOT-0402-25G COBBOT-0402-1G
4-(Methylsulphinyl)phenol
4-(Methylsulphinyl)phenol
Formula: C₇H₈O₂S
MDL Number: MFCD00464311
CAS Number: 14763-64-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR