4-Nitrophenyl phenyl sulphide

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

002980-5G 002980-25G
FLUO002980-5GEA 0 GBP
FLUO002980-5G FLUO002980-25G
4-Nitrophenyl phenyl sulphide
4-Nitrophenyl phenyl sulphide
Formula: C₁₂H₉NO₂S
MDL Number: MFCD00024700
CAS Number: 952-97-6
EINECS: 213-462-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR