N,N-Dimethyl-4-fluoro-2-nitroaniline
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
4-Fluoro-N,N-dimethyl-2-nitroaniline, N,N-Dimethyl 4-fluoro-2-nitroaniline
AN-1224-1G
AN-1224-5G
AN-1224-25G
COBBAN-1224-1GEA
0
GBP
COBBAN-1224-1G
COBBAN-1224-5G
COBBAN-1224-25G
N,N-Dimethyl-4-fluoro-2-nitroaniline
N,N-Dimethyl-4-fluoro-2-nitroaniline
Formula:
C₈H₉FN₂O₂ MW: 184.17 g/mol |
MDL Number:
MFCD04116334 CAS Number: 69261-01-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...