5-Carboxy-2-fluorophenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Borono-4-fluorobenzoic acid

BB-2883-5G BB-2883-1G
COBBBB-2883-5GEA 0 GBP
COBBBB-2883-5G COBBBB-2883-1G
5-Carboxy-2-fluorophenylboronic acid
5-Carboxy-2-fluorophenylboronic acid
Formula: C₇H₆BFO₄
MW: 183.9300 g/mol
MDL Number: MFCD08436065
CAS Number: 874219-59-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR