3-Carboxy-4-fluorophenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Carboxy-4-fluorophenylboronic Acid, 3-Carboxy-4-fluorobenzeneboronic acid

BB-2879-25G BB-2879-1G BB-2879-5G
COBBBB-2879-25GEA 0 GBP
COBBBB-2879-25G COBBBB-2879-1G COBBBB-2879-5G
3-Carboxy-4-fluorophenylboronic acid
3-Carboxy-4-fluorophenylboronic acid
Formula: C₇H₆BFO₄
MW: 183.93 g/mol
MDL Number: MFCD03095129
CAS Number: 872460-12-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR