5-(Ethoxycarbonyl)-2-fluorophenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (5-(Ethoxycarbonyl)-2-fluorophenyl)boronic acid, 5-Ethoxycarbonyl-2-fluorobenzeneboronic acid

BB-2885-5G BB-2885-1G
COBBBB-2885-5GEA 0 GBP
COBBBB-2885-5G COBBBB-2885-1G
5-(Ethoxycarbonyl)-2-fluorophenylboronic acid
5-(Ethoxycarbonyl)-2-fluorophenylboronic acid
Formula: C₉H₁₀BFO₄
MW: 211.98 g/mol
MDL Number: MFCD08235094
CAS Number: 874219-60-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR