3-(Ethoxycarbonyl)-4-fluorophenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Ethoxycarbonyl-4-fluorophenylboronic acid, (3-Ethoxycarbonyl-4-fluorophenyl)boronic acid

BB-2861-5G BB-2861-1G
COBBBB-2861-5GEA 0 GBP
COBBBB-2861-5G COBBBB-2861-1G
3-(Ethoxycarbonyl)-4-fluorophenylboronic acid
3-(Ethoxycarbonyl)-4-fluorophenylboronic acid
Formula: C₉H₁₀BFO₄
MW: 211.9827 g/mol
MDL Number: MFCD08061836
CAS Number: 874219-36-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR