4-Fluoro-3-(methylcarbamoyl)phenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (4-Fluoro-3-(methylcarbamoyl)phenyl)boronic acid, 4-Fluoro-3-(methylcarbamoyl)benzeneboronic acid

BB-2862-5G BB-2862-1G
COBBBB-2862-5GEA 0 GBP
COBBBB-2862-5G COBBBB-2862-1G
4-Fluoro-3-(methylcarbamoyl)phenylboronic acid
4-Fluoro-3-(methylcarbamoyl)phenylboronic acid
Formula: C₈H₉BFNO₃
MDL Number: MFCD08235038
CAS Number: 874219-19-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR