3-(Dimethylcarbamoyl)-5-fluorophenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BB-2853-5G BB-2853-1G
COBBBB-2853-5GEA 0 GBP
COBBBB-2853-5G COBBBB-2853-1G
3-(Dimethylcarbamoyl)-5-fluorophenylboronic acid
3-(Dimethylcarbamoyl)-5-fluorophenylboronic acid
Formula: C₉H₁₁BFNO₃
MDL Number: MFCD08235056
CAS Number: 874219-39-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR