3-(Dimethylcarbamoyl)-4-fluorophenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (3-(Dimethylcarbamoyl)-4-fluorophenyl)boronic acid, 3-Dimethylcarbamoyl-4-fluorobenzeneboronic acid, 4-Fluoro-3-(dimethylcarbamoyl)phenylboronic acid

BB-2870-5G BB-2870-1G
COBBBB-2870-5GEA 0 GBP
COBBBB-2870-5G COBBBB-2870-1G
3-(Dimethylcarbamoyl)-4-fluorophenylboronic acid
3-(Dimethylcarbamoyl)-4-fluorophenylboronic acid
Formula: C₉H₁₁BFNO₃
MDL Number: MFCD08235046
CAS Number: 874219-27-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR