2-Chloro-(5-methoxycarbonyl)phenylboronic acid ≥98%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
2-Chloro-5-(methoxycarbonyl)phenylboronic acid, (2-Chloro-5-(methoxycarbonyl)phenyl)boronic acid, 2-Chloro-5-(methoxycarbonyl)benzeneboronic acid
H52999.MD
H52999.03
H52999.MDEA
0
GBP
H52999.MD
H52999.03
2-Chloro-(5-methoxycarbonyl)phenylboronic acid ≥98%
2-Chloro-(5-methoxycarbonyl)phenylboronic acid
Purity: 98%
CAS No.: 913835-92-4
Molecular Formula: C8H8BClO4
Formula Weight: 214.41
Melting Point: 230-232°
MDL No.: MFCD08689528
Formula:
C₈H₈BClO₄ MW: 214.41 g/mol Melting Pt: 230…232 °C Storage Temperature: Ambient |
MDL Number:
MFCD08689528 CAS Number: 913835-92-4 |
Learn more
![](/m-uk.vwr.com/en_GB/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...