4-Bromo-2-fluoro-5-nitroaniline
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AN-1533-1G
AN-1533-5G
COBBAN-1533-1GEA
0
GBP
COBBAN-1533-1G
COBBAN-1533-5G
4-Bromo-2-fluoro-5-nitroaniline
4-Bromo-2-fluoro-5-nitroaniline
Formula:
C₆H₄BrFN₂O₂ MW: 235.0106 g/mol |
MDL Number:
MFCD08741367 CAS Number: 87547-06-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...