3-Carboxyphenylboronic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Boronobenzoic acid, 3-Carboxybenzeneboronic acid

010785-25G 010785-5G
FLUO010785-25GEA 0 GBP
FLUO010785-25G FLUO010785-5G
3-Carboxyphenylboronic acid
3-Carboxyphenylboronic acid
Formula: C₇H₇BO₄
MW: 165.94 g/mol
MDL Number: MFCD00036833
CAS Number: 25487-66-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR