3-(2,3-Dimethylphenylcarbamoyl)phenylboronic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BB-3857-5G
BB-3857-1G
COBBBB-3857-5GEA
0
GBP
COBBBB-3857-5G
COBBBB-3857-1G
3-(2,3-Dimethylphenylcarbamoyl)phenylboronic acid
3-(2,3-Dimethylphenylcarbamoyl)phenylboronic acid
Formula:
C₁₅H₁₆BNO₃ |
MDL Number:
MFCD09027262 CAS Number: 957060-99-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...