3-Carboxyphenylboronic acid pinacol ester

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Carboxybenzeneboronic acid pinacol ester, 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid

BB-2002-25G BB-2002-100G BB-2002-1G BB-2002-5G
COBBBB-2002-25GEA 0 GBP
COBBBB-2002-25G COBBBB-2002-100G COBBBB-2002-1G COBBBB-2002-5G
3-Carboxyphenylboronic acid pinacol ester
3-Carboxyphenylboronic acid pinacol ester
Formula: C₁₃H₁₇BO₄
MW: 248.083 g/mol
MDL Number: MFCD03411930
CAS Number: 269409-73-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR