3-(Ethoxycarbonyl)phenylboronic acid pinacol ester

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Ethyl 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate, 3-Ethoxycarbonylphenylboronic acid, pinacol ester, Ethyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

PN-2117-5G PN-2117-1G
COBBPN-2117-5GEA 0 GBP
COBBPN-2117-5G COBBPN-2117-1G
3-(Ethoxycarbonyl)phenylboronic acid pinacol ester
3-(Ethoxycarbonyl)phenylboronic acid pinacol ester
Formula: C₁₅H₂₁BO₄
MW: 276.136 g/mol
MDL Number: MFCD03093896
CAS Number: 269410-00-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR