2-(Ethoxycarbonyl)phenylboronic acid pinacol ester

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Ethoxycarbonylphenylboronic acid pinacol ester, 2-(Ethoxycarbonyl)benzeneboronic acid pinacol ester, Ethyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate, Ethyl 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

PN-2126-5G PN-2126-1G
COBBPN-2126-5GEA 0 GBP
COBBPN-2126-5G COBBPN-2126-1G
2-(Ethoxycarbonyl)phenylboronic acid pinacol ester
2-(Ethoxycarbonyl)phenylboronic acid pinacol ester
Formula: C₁₅H₂₁BO₄
MW: 276.136 g/mol
MDL Number: MFCD03453660
CAS Number: 269409-99-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR