2-Amino-4-bromo-6-fluorobenzothiazole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Bromo-6-fluorobenzo[d]thiazol-2-amine

HC-2118-5G HC-2118-1G
COBBHC-2118-5GEA 0 GBP
COBBHC-2118-5G COBBHC-2118-1G
2-Amino-4-bromo-6-fluorobenzothiazole
2-Amino-4-bromo-6-fluorobenzothiazole
Formula: C₇H₄BrFN₂S
MW: 247.0875 g/mol
MDL Number: MFCD02664333
CAS Number: 383131-15-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR