Hexafluoroantimonic acid hexahydrate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

006553-200G 006553-500G
FLUO006553-200GEA 0 GBP
FLUO006553-200G FLUO006553-500G
Hexafluoroantimonic acid hexahydrate
Hexafluoroantimonic acid hexahydrate
Formula: HF₆Sb·6H₂O
MDL Number: MFCD00149905
CAS Number: 72121-43-8
EINECS: 241-023-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR