1,5-Dimethyl-1H-pyrazole-3-carboxylic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CFF459.2.5 CFF459.5 CFF459.10 017472-2.5G 017472-5G 017472-10G 017472-1G
CHMPCFF459.2.5EA 0 GBP
CHMPCFF459.2.5 CHMPCFF459.5 CHMPCFF459.10 FLUO017472-2.5G FLUO017472-5G FLUO017472-10G FLUO017472-1G
1,5-Dimethyl-1H-pyrazole-3-carboxylic acid
1,5-Dimethyl-1H-pyrazole-3-carboxylic acid
Formula: C₆H₈N₂O₂
MW: 140.14 g/mol
MDL Number: MFCD00085040
CAS Number: 5744-59-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR