1,3-Dimethyl-1H-pyrazole-5-carbonyl chloride
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CFF458.2.5
CFF458.5
CFF458.10
017471-2.5G
017471-5G
017471-10G
017471-1G
CHMPCFF458.2.5EA
0
GBP
CHMPCFF458.2.5
CHMPCFF458.5
CHMPCFF458.10
FLUO017471-2.5G
FLUO017471-5G
FLUO017471-10G
FLUO017471-1G
1,3-Dimethyl-1H-pyrazole-5-carbonyl chloride
1,3-Dimethyl-1H-pyrazole-5-carbonyl chloride
Formula:
C₆H₇ClN₂O |
MDL Number:
MFCD00833258 CAS Number: 55458-67-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...