Tetrahydropyran-4-methanol

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS20467G1 AS20467G25 AS20467G5
ACTIAS20467G1EA 0 GBP
ACTIAS20467G1 ACTIAS20467G25 ACTIAS20467G5
Tetrahydropyran-4-methanol
Tetrahydropyran-4-methanol
Formula: C₆H₁₂O₂
MDL Number: MFCD00457804
CAS Number: 14774-37-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR