Tetrahydropyran-2-methanol

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AM-1020-5G AM-1020-100G AM-1020-25G
COBBAM-1020-5GEA 0 GBP
COBBAM-1020-5G COBBAM-1020-100G COBBAM-1020-25G
Tetrahydropyran-2-methanol
Tetrahydropyran-2-methanol
Formula: C₆H₁₂O₂
MDL Number: MFCD00006624
CAS Number: 100-72-1
EINECS: 202-882-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR