5-Indoleboronic acid
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
(1H-indol-5-yl)boronic acid, (1H-Indol-5-yl)boronic acid, 5-Indolylboronic acid
011040-5G
011040-1G
FLUO011040-5GEA
0
GBP
FLUO011040-5G
FLUO011040-1G
5-Indoleboronic acid
5-Indoleboronic acid
Formula:
C₈H₈BNO₂ MW: 160.97 g/mol |
MDL Number:
MFCD01319013 CAS Number: 144104-59-6 |
Learn more
![](/m-uk.vwr.com/en_GB/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...