5-Indoleboronic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
(1H-indol-5-yl)boronic acid, (1H-Indol-5-yl)boronic acid, 5-Indolylboronic acid
BB-2319-5G
BB-2319-1G
COBBBB-2319-5GEA
0
GBP
COBBBB-2319-5G
COBBBB-2319-1G
5-Indoleboronic acid
5-Indoleboronic acid
Formula:
C₈H₈BNO₂ MW: 160.97 g/mol |
MDL Number:
MFCD01319013 CAS Number: 144104-59-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...