4-Indoleboronic acid
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
4-Indolylboronic acid, (1H-Indol-4-yl)boronic acid
016109-1G
016109-5G
FLUO016109-1GEA
0
GBP
FLUO016109-1G
FLUO016109-5G
4-Indoleboronic acid
4-Indoleboronic acid
Formula:
C₈H₈BNO₂ MW: 160.97 g/mol |
MDL Number:
MFCD03095175 CAS Number: 220465-43-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...