2-Methoxy-4-(trifluoromethyl)phenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (2-Methoxy-4-(trifluoromethyl)phenyl)boronic acid

BB-5302-1G BB-5302-5G
COBBBB-5302-1GEA 0 GBP
COBBBB-5302-1G COBBBB-5302-5G
2-Methoxy-4-(trifluoromethyl)phenylboronic acid
2-Methoxy-4-(trifluoromethyl)phenylboronic acid
Formula: C₈H₈BF₃O₃
MW: 219.9500 g/mol
MDL Number: MFCD03428511
CAS Number: 312936-89-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR