5-Chloro-2-methylphenylboronic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BB-2208-1G
BB-2208-5G
BB-2208-25G
COBBBB-2208-1GEA
0
GBP
COBBBB-2208-1G
COBBBB-2208-5G
COBBBB-2208-25G
5-Chloro-2-methylphenylboronic acid
5-Chloro-2-methylphenylboronic acid
Formula:
C₇H₈BClO₂ |
MDL Number:
MFCD03411939 CAS Number: 148839-33-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...