3-Chloro-2-methylphenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BB-2035-5G BB-2035-25G BB-2035-1G
COBBBB-2035-5GEA 0 GBP
COBBBB-2035-5G COBBBB-2035-25G COBBBB-2035-1G
3-Chloro-2-methylphenylboronic acid
3-Chloro-2-methylphenylboronic acid
Formula: C₇H₈BClO₂
MDL Number: MFCD04115641
CAS Number: 313545-20-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR