2-Chloro-5-methylphenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (2-Chloro-5-methylphenyl)boronic acid, 2-Chloro-5-methylbenzeneboronic acid

BB-2180-5G BB-2180-25G BB-2180-1G
COBBBB-2180-5GEA 0 GBP
COBBBB-2180-5G COBBBB-2180-25G COBBBB-2180-1G
2-Chloro-5-methylphenylboronic acid
2-Chloro-5-methylphenylboronic acid
Formula: C₇H₈BClO₂
MW: 170.40 g/mol
MDL Number: MFCD06659822
CAS Number: 193353-35-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR