Ethyl-3-hydroxyhexanoate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Ethyl 3-hydroxyhexanoate

M-Clarity™ quality level = MQ300

W354503-100G-K W354503-1KG-K W354503-5KG-K
SIALW354503-100G-KEA 139 GBP
SIALW354503-100G-K SIALW354503-1KG-K SIALW354503-5KG-K
Ethyl-3-hydroxyhexanoate, Sigma-Aldrich®
Ethyl 3-hydroxyhexanoate
Formula: C₈H₁₆O₃
MW: 160.21 g/mol
MDL Number: MFCD00036604
CAS Number: 2305-25-1
EINECS: 218-973-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR