2-Cyano-4'-fluoroacetanilide

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS23642G25 AS23642G5
ACTIAS23642G25EA 0 GBP
ACTIAS23642G25 ACTIAS23642G5
2-Cyano-4'-fluoroacetanilide
2-Cyano-4'-fluoroacetanilide
Formula: C₉H₇FN₂O
MDL Number: MFCD00276110
CAS Number: 1735-88-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR