2-Amino-N-methylbenzamide

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21688G100 AS21688G25 AS21688G5
ACTIAS21688G100EA 218 GBP
ACTIAS21688G100 ACTIAS21688G25 ACTIAS21688G5
2-Amino-N-methylbenzamide
2-Amino-N-methylbenzamide
Formula: C₈H₁₀N₂O
MW: 150.1778 g/mol
MDL Number: MFCD00060602
CAS Number: 4141-08-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR