2-Fluoro-3-nitrotoluene

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Fluoro-1-methyl-3-nitrobenzene

AS21860G100 AS21860G25
ACTIAS21860G100EA 0 GBP
ACTIAS21860G100 ACTIAS21860G25
2-Fluoro-3-nitrotoluene
2-Fluoro-3-nitrotoluene
Formula: C₇H₆FNO₂
MDL Number: MFCD03412242
CAS Number: 437-86-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR