3-Bromo-2-fluorotoluene

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS23059G100 AS23059G25 AS23059G5
ACTIAS23059G100EA 0 GBP
ACTIAS23059G100 ACTIAS23059G25 ACTIAS23059G5
3-Bromo-2-fluorotoluene
3-Bromo-2-fluorotoluene
Formula: C₇H₆BrF
MDL Number: MFCD04972694
CAS Number: 59907-12-9
EINECS: 261-981-0

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR