2,5-Difluoro-4-nitrobenzoic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS22692G1 AS22692G25 AS22692G5
ACTIAS22692G1EA 0 GBP
ACTIAS22692G1 ACTIAS22692G25 ACTIAS22692G5
2,5-Difluoro-4-nitrobenzoic acid
2,5-Difluoro-4-nitrobenzoic acid
Formula: C₇H₃F₂NO₄
MW: 203.10 g/mol
MDL Number: MFCD03425705
CAS Number: 116465-48-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR