4-Chloro-2-fluoro-5-nitrobenzoic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21640G25 AS21640G5
ACTIAS21640G25EA 0 GBP
ACTIAS21640G25 ACTIAS21640G5
4-Chloro-2-fluoro-5-nitrobenzoic acid
4-Chloro-2-fluoro-5-nitrobenzoic acid
Formula: C₇H₃ClFNO
MDL Number: MFCD04115725
CAS Number: 35112-05-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR