3-Bromo-2-methylthiophene

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS33769G1 AS33769G5
ACTIAS33769G1EA 47 GBP
ACTIAS33769G1 ACTIAS33769G5
3-Bromo-2-methylthiophene
3-Bromo-2-methylthiophene
Formula: C₅H₅BrS
MW: 177.0622 g/mol
MDL Number: MFCD09033008
CAS Number: 30319-05-2
EINECS: 250-128-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR